Merck
  • Parallel solid-phase synthesis of trisubstituted triazinobenzimidazolediones.
Parallel solid-phase synthesis of trisubstituted triazinobenzimidazolediones.
Journal of combinatorial chemistry (2002-07-09)
Gérard Klein, Achyuta N Acharya, John M Ostresh, Richard A Houghten
ABSTRACT
An efficient method for the solid-phase synthesis of trisubstituted [1,3,5]triazino[1,2-a]benzimidazole-2,4(3H,10H)-diones from resin-bound amino acids is described. N-acylation of the primary amine of a resin-bound amino acid with 4-fluoro-3-nitrobenzoic acid, followed by displacement of the fluoro group and reduction of the nitro group, generated a resin-bound o-dianilino derivative. The dianilino compound was treated with cyanogen bromide to generate the corresponding iminobenzimidazole, which, following treatment with N-(chlorocarbonyl)isocyanate, afforded the resin-bound triazinodione derivative. Alkylation of the triazinodione compound with an alkyl halide yielded, following cleavage of the solid-support, the trisubstituted [1,3,5]triazino[1,2-a]benzimidazole-2,4(3H,10H)-dione.
Sigma-Aldrich
4-Fluoro-3-nitrobenzoic acid, 98%
Pricing and availability is not currently available.