Merck
The anti-cancer activity of noscapine: a review.
Recent patents on anti-cancer drug discovery (2009-01-20)
Massoud Mahmoudian, Parvaneh Rahimi-Moghaddam
ABSTRACT
Noscapine is an isoqiunoline alkaloid found in opium latex. Unlike most other alkaloids obtained from opium latex, noscapine is not sedative and has been used as antitussive drug in various countries. Recently, it has been introduced as an anti-mitotic agent. This drug can be used orally. When the resistance to other anti-cancer drugs such as paclitaxel manifests, noscapine might be effective. Therefore, noscapine and its analogs have great potential as novel anti-cancer agents.
Product Description
Noscapine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
(S,R)-Noscapine, 97%
Pricing and availability is not currently available.