Merck
Michael reactions carried out using a bench-top flow system.
Organic & biomolecular chemistry (2006-02-01)
Frederic Bonfils, Isabelle Cazaux, Philip Hodge, Claude Caze
ABSTRACT
The Michael reaction between methyl 1-oxoindan-2-carboxylate and methyl vinyl ketone was achieved successfully by pumping solutions of the reactants in toluene through a fluid bed of Amberlyst A21 at 50 degrees C. The use of a fluid bed reactor is attractive as it allows gel-type beads, i.e. the type of bead used in most studies of polymer-supported (PS) organic reactions, to be used satisfactorily in a flow system. When polymer-supported cinchonidine was used in place of Amberlyst A21, the Michael product was obtained in high yield with an enantiomeric excess (ee) of 51%. This % ee is comparable to that achieved when the reaction was catalysed by cinchonidine itself.
Product Description
Supelco
Amberlyst® A21 free base, free base
Pricing and availability is not currently available.