Merck
  • Finite element analysis of provisional structures of implant-supported complete prostheses.
Finite element analysis of provisional structures of implant-supported complete prostheses.
The Journal of oral implantology (2014-05-02)
Bruno Albuquerque Carneiro, Rui Barbosa de Brito, Fabiana Mantovani Gomes França
ABSTRACT
The use of provisional resin implant-supported complete dentures is a fast and safe procedure to restore mastication and esthetics of patients soon after surgery and during the adaptation phase to the new denture. This study assessed stress distribution of provisional implant-supported fixed dentures and the all-on-4 concept using self-curing acrylic resin (Tempron) and bis-acrylic resin (Luxatemp) to simulate functional loads through the three-dimensional finite element method. Solidworks software was used to build three-dimensional models using acrylic resin (Tempron, model A) and bis-acrylic resin (Luxatemp, model B) for denture captions. Two loading patterns were applied on each model: (1) right unilateral axial loading of 150 N on the occlusal surfaces of posterior teeth and (2) oblique loading vector of 150 N at 45°. The results showed that higher stress was found on the bone crest below oblique load application with a maximum value of 187.57 MPa on model A and 167.45 MPa on model B. It was concluded that model B improved stress distribution on the denture compared with model A.
Sigma-Aldrich
Bisphenol A glycerolate dimethacrylate, glycerol/phenol 1
Pricing and availability is not currently available.