Merck
  • miR-223 regulates cell growth and targets proto-oncogenes in mycosis fungoides/cutaneous T-cell lymphoma.
miR-223 regulates cell growth and targets proto-oncogenes in mycosis fungoides/cutaneous T-cell lymphoma.
The Journal of investigative dermatology (2013-12-07)
Laura Y McGirt, Clare M Adams, Devin A Baerenwald, Jeffrey P Zwerner, John A Zic, Christine M Eischen
ABSTRACT
The pathogenesis of the cutaneous T-cell lymphoma (CTCL), mycosis fungoides (MF), is unclear. MicroRNA (miRNA) are small noncoding RNAs that target mRNA leading to reduced mRNA translation. Recently, specific miRNA were shown to be altered in CTCL. We detected significantly reduced expression of miR-223 in early-stage MF skin, and further decreased levels of miR-223 in advanced-stage disease. CTCL peripheral blood mononuclear cells and cell lines also had reduced miR-223 as compared with controls. Elevated expression of miR-223 in these cell lines reduced cell growth and clonogenic potential, whereas inhibition of miR-223 increased cell numbers. Investigations into putative miR-223 targets with oncogenic function, including E2F1 and MEF2C, and the predicted miR-223 target, TOX, revealed that all three were targeted by miR-223 in CTCL. E2F1, MEF2C, and TOX proteins were decreased with miR-223 overexpression, whereas miR-223 inhibition led to increased protein levels in CTCL. In addition, we showed that the 3'-UTR of TOX mRNA was a genuine target of miR-223. Therefore, reduced levels of miR-223 in MF/CTCL lead to increased expression of E2F1, MEF2C, and TOX, which likely contributes to the development and/or progression of CTCL. Thus, miR-223 and its targets may be useful for the development of new therapeutics for MF/CTCL.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Pricing and availability is not currently available.