Merck
  • Behavioural changes in children following minor surgery--is premedication beneficial?
Behavioural changes in children following minor surgery--is premedication beneficial?
Acta anaesthesiologica Belgica (1992-01-01)
K A Payne, A R Coetzee, F J Mattheyse, J J Heydenrych
ABSTRACT
One hundred and twenty-three male children, aged one to ten years, were studied to determine the influence of premedication on changes in patterns of behaviour following hospitalization for repair of inguinal hernias. Four comparable groups were selected for premedication regimen: (1) A control group without premedication; (2) oral trimeprazine tartrate 2 mg/kg, methadone 0.1 mg/kg and droperidol 0.15 mg/kg; (3) oral midazolam 0.45 mg/kg; (4) intramuscular midazolam 0.15 mg/kg. Standard inhalational anesthesia was used and caudal blocks employed for analgesia. The parents returned a questionnaire at two weeks. Changes in behaviour were reported in 78% of the children and overall, premedication showed little benefit. However, midazolam premedication was associated with a significantly lower incidence of night-time crying and awakening, compared with no premedication. Only for night-time crying and day-time toilet training did age below five years prove to be a significant contributing factor.
Product Description
Alimemazine hemitartrate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Alimemazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.