Merck
  • Intramolecular direct C-H arylation approach to fused purines. Synthesis of purino[8,9-f]phenanthridines and 5,6-dihydropurino[8,9-a]isoquinolines.
Intramolecular direct C-H arylation approach to fused purines. Synthesis of purino[8,9-f]phenanthridines and 5,6-dihydropurino[8,9-a]isoquinolines.
The Journal of organic chemistry (2010-03-05)
Igor Cerna, Radek Pohl, Blanka Klepetárová, Michal Hocek
ABSTRACT
Intramolecular C-H arylations were employed as a key step in the synthesis of hitherto unknown fused purine systems: 13-substituted purino[8,9-f]phenanthridines and 11-substituted 5,6-dihydropurino[8,9-a]isoquinolines. The purino[8,9-f]phenanthridines were prepared in moderate yields by double C-H arylations of 9-phenylpurines with 1,2-diiodobenzene or, more efficiently, by consecutive Suzuki coupling of 9-(2-bromophenyl)purines with 2-bromophenylboronic acid followed by intramolecular C-H arylation. 5,6-Dihydropurino[8,9-a]isoquinolines were prepared in quantitative yields by intramolecular C-H arylations of 9-(2-chlorophenethyl)purines.
Sigma-Aldrich
1,2-Diiodobenzene, 98%
Pricing and availability is not currently available.