Merck
  • Activity of MK-7655 combined with imipenem against Enterobacteriaceae and Pseudomonas aeruginosa.
Activity of MK-7655 combined with imipenem against Enterobacteriaceae and Pseudomonas aeruginosa.
The Journal of antimicrobial chemotherapy (2013-05-23)
David M Livermore, Marina Warner, Shazad Mushtaq
ABSTRACT
MK-7655 is a novel inhibitor of class A and C β-lactamases. We investigated its potential to protect imipenem. Chequerboard MICs were determined by CLSI agar dilution: (i) for Enterobacteriaceae with carbapenemases; (ii) for Enterobacteriaceae with carbapenem resistance contingent on combinations of impermeability together with an extended-spectrum β-lactamase or AmpC enzyme; and (iii) for Pseudomonas aeruginosa and other non-fermenters. At a concentration of 4 mg/L, MK-7655 reduced imipenem MICs for Enterobacteriaceae with KPC carbapenemases from 16-64 mg/L to 0.12-1 mg/L. Synergy also was seen for Enterobacteriaceae with impermeability-mediated carbapenem resistance, with weaker synergy seen for isolates with the OXA-48 enzyme. On the other hand, MK-7655 failed to potentiate imipenem against Enterobacteriaceae with metallo-carbapenemases. In the case of P. aeruginosa, where endogenous AmpC confers slight protection versus imipenem, 4 mg/L MK-7655 reduced the MIC of imipenem for all isolates, except those with metallo-carbapenemases: the MICs of imipenem fell from 1-2 mg/L to 0.25-0.5 mg/L for imipenem-susceptible P. aeruginosa and from 16-64 mg/L to 1-4 mg/L for OprD-deficient strains. No potentiation was seen for chryseobacteria or for Stenotrophomonas maltophilia. MK-7655 potentiated imipenem against Enterobacteriaceae with KPC carbapenemases or combinations of β-lactamase and impermeability, but not those with metallo-carbapenemases. It augmented the activity of imipenem against P. aeruginosa in general and OprD mutants in particular.
Sigma-Aldrich
Imipenem monohydrate, ≥98% (HPLC)
Pricing and availability is not currently available.