Merck
  • Organocatalytic C3-selective Friedel-Crafts alkylations of indoles with alpha,beta-unsaturated ketones.
Organocatalytic C3-selective Friedel-Crafts alkylations of indoles with alpha,beta-unsaturated ketones.
Chemical communications (Cambridge, England) (2006-02-09)
Dong-Ping Li, Ying-Cen Guo, Yu Ding, Wen-Jing Xiao
ABSTRACT
The use of an equimolar amount of pyrrolidine and HClO4 (30 mol%) was found to be effective in promoting the conjugate addition of indoles to (E)-alpha,beta-unsaturated ketones, affording the corresponding beta-indolyl ketones in excellent yields.
Sigma-Aldrich
5-Methyl-3-hexen-2-one, technical grade, 75%
Pricing and availability is not currently available.