Merck
  • Predominance of spliceosomal complex formation over polyadenylation site selection in TDP-43 autoregulation.
Predominance of spliceosomal complex formation over polyadenylation site selection in TDP-43 autoregulation.
Nucleic acids research (2013-12-27)
Sara Bembich, Jeremias S Herzog, Laura De Conti, Cristiana Stuani, S Eréndira Avendaño-Vázquez, Emanuele Buratti, Marco Baralle, Francisco E Baralle
ABSTRACT
TDP-43 is a nuclear protein involved in many aspects of RNA metabolism. To ensure cellular viability, its expression levels within cells must be tightly regulated. We have previously demonstrated that TDP-43 autoregulation occurs through the activation of a normally silent intron in its 3'-UTR sequence that results in the use of alternative polyadenylation sites. In this work, we analyse which is the dominant event in autoregulation: the recognition of the splice sites of 3'-UTR intron 7 or the intrinsic quality of the alternative polyadenylation sites. A panel of minigene constructs was tested for autoregulation functionality, protein production and subcellular messenger RNA localization. Our data clearly indicate that constitutive spliceosome complex formation across intron 7 does not lead to high protein production but, on the contrary, to lower TDP-43 messenger RNA and protein levels. This is due to altered nucleocytoplasmic distribution of the RNA that is mostly retained in the nucleus and degraded. This study provides a novel in-depth characterization of how RNA binding proteins can autoregulate their own levels within cells, an essential regulatory process in maintaining cellular viability.
Product Description
Roche
Anti-Digoxigenin-Rhodamine, Fab fragments, from sheep
Pricing and availability is not currently available.
Roche
Digoxigenin-11-dUTP, alkali-labile, =85% (HPLC), solution, pkg of 25 μL (11573152910 [1 mM]), pkg of 125 μL (11573179910 [1 mM]]])
Pricing and availability is not currently available.