Merck
  • Cobalt-catalyzed cross-coupling between in situ prepared arylzinc halides and 2-chloropyrimidine or 2-chloropyrazine.
Cobalt-catalyzed cross-coupling between in situ prepared arylzinc halides and 2-chloropyrimidine or 2-chloropyrazine.
The Journal of organic chemistry (2009-03-25)
Jeanne-Marie Bégouin, Corinne Gosmini
ABSTRACT
A cobalt-catalyzed cross-coupling of aryl halides with 2-chloropyrimidines or 2-chloropyrazines is reported in satisfactory to high yields. The key step of this procedure is the formation of aromatic organozinc reagents and their coupling with 2-chlorodiazines using the same cobalt halide as catalyst and Zn dust under mild reaction conditions. This new cobalt-catalyzed coupling reaction represents a practical and interesting alternative to previously known methods for the synthesis of 2-aryldiazines.
Sigma-Aldrich
2-Chloropyrazine, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Chloropyrimidine, 95%
Pricing and availability is not currently available.