Merck
  • Pattern of aliphatic dicarboxylic acids in uremic serum including a new organic acid, 2,4-dimethyladipic acid.
Pattern of aliphatic dicarboxylic acids in uremic serum including a new organic acid, 2,4-dimethyladipic acid.
Clinica chimica acta; international journal of clinical chemistry (1979-11-15)
T Niwa, T Ohki, K Maeda, A Saito, K Kobayashi
PMID498544
ABSTRACT
(1) 2,4-Dimethyladipic acid was first identified in normal human urine using gas chromatography-mass spectrometry. Urinary excretion of 2,4-dimethyladipic acid in 7 healthy adults ranged from 4.9 mumol to 14 mumol per 24 h. (2) Succinic acid, adipic acid, 3-methyladipic acid, 2,4-dimethyladipic acid, pimelic acid and azelaic acid were identified in the ultrafiltrate of the blood obtained from a chronic uremic patient using a hemodialyzer. (3) Levels of succinic acid, adipic acid, 3-methyladipic acid, 2,4-dimethyladipic acid, pimelic acid and azelaic acid in uremic serum were determined using a mass fragmentographic technique. Concentration of succinic acid in uremic serum was comparable to that in normal serum, whereas concentrations of adipic acid, 3-methyladipic acid, 2,4-dimethyladipic acid, pimelic acid and azelaic acid were highly elevated in uremic serum.
Sigma-Aldrich
Pimelic acid, 98%
Pricing and availability is not currently available.