Merck
  • Prolonged protection against exercise-induced bronchoconstriction by the leukotriene D4-receptor antagonist cinalukast.
Prolonged protection against exercise-induced bronchoconstriction by the leukotriene D4-receptor antagonist cinalukast.
The Journal of allergy and clinical immunology (1997-02-01)
E Adelroth, M D Inman, E Summers, D Pace, M Modi, P M O'Byrne
ABSTRACT
The degree and duration of protection against exercise-induced bronchoconstriction afforded by three doses of a specific leukotriene D4 receptor antagonist, cinalukast, were assessed after an initial dosing and after 1 week of therapy. A placebo-controlled crossover study was performed in eight male patients who had mild, stable asthma and exercise-induced bronchoconstriction. Treatment consisted of four 7-day periods of placebo and three dose levels of the drug (10, 50, and 200 mg administered orally). Exercise challenge was performed at 2 hours and 8 hours after treatment on the first and seventh treatment days. The response was measured as the area under the FEV1-time effect curve (AUEC). On the first day of treatment, the mean (+/- SEM) AUEC at 2 hours was 24.2 +/- 3.3 L.min after placebo and was 5.5 +/- 2.2 L.min, 6.3 +/- 2.7 L.min, 3.3 +/- 3.8 L.min after 10 mg, 50 mg, and 200 mg, respectively (p < 0.05 for all values compared with placebo). The AUEC at 8 hours on the first day was 25.1 +/- 4.4 L.min after placebo and was 6.8 +/- 4.1 L.min, 11.2 +/- 2.5 L.min, and 5.0 +/- 2.8 L.min after 10 mg, 50 mg, and 200 mg, respectively (p < 0.05 for all values compared with placebo). The protection afforded by 10 mg of cinaluicast was lost after 7 days of treatment but persisted with 50 mg and 200 mg doses. Orally administered cinalukast provides at least 8 hours of protection against exercise-induced bronchoconstriction. This protection is lost with regular treatment for 1 week for the lowest dose studied.
Sigma-Aldrich
Cinalukast, ~98% (HPLC)
Pricing and availability is not currently available.