Merck
  • Catalysis of the hydrolysis of ethyl mandelate and esterification of alpha-bromopropionic acid by lipase in microemulsions.
Catalysis of the hydrolysis of ethyl mandelate and esterification of alpha-bromopropionic acid by lipase in microemulsions.
Chinese journal of biotechnology (1993-01-01)
H Xiao, J Liu, Z Li
ABSTRACT
Candida cyclindracea lipase (CCL) was added to "sodium dodecyl sulfonate (AS)/n-butanol/n-octane/n-octane" water-in-oil microemulsion to catalyze the hydrolysis of ethyl mandelate and the esterification of alpha-bromopropionic acid with n-butanol, respectively. The catalytic activity of CCL in the above microemulsions was higher than that in the traditional oil/water biphasic systems. After hydrolysis for 48 h, the conversion rate of the reaction reached 90% and S-mandelic acid, [alpha]D20-149.8 (C10; H2O), optical purity ca. 97%, was isolated. While after esterification for 6 h, the conversion rate of the reaction reached 45%, and butyl-(R)-alpha-bromopropionate, [alpha]D20 18.2 (Cl; CHCl3), optical purity ca. 99%, was obtained.
Sigma-Aldrich
2-Bromopropionic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl (R)-(−)-mandelate, 99%, optical purity ee: 99% (GLC)
Pricing and availability is not currently available.