Merck
  • Characterization of the safener-induced glutathione S-transferase isoform II from maize.
Characterization of the safener-induced glutathione S-transferase isoform II from maize.
Planta (1995-01-01)
D C Holt, V J Lay, E D Clarke, A Dinsmore, I Jepson, S W Bright, A J Greenland
ABSTRACT
The safener-induced maize (Zea mays L.) glutathione S-transferase, GST II (EC 2.5.1.18) and another predominant isoform, GST I, were purified from extracts of maize roots treated with the safeners R-25788 (N,N-diallyl-2-dichloroacetamide) or R-29148 (3-dichloroacetyl-2,2,5-trimethyl-1,3-oxazolidone). The isoforms GST I and GST II are respectively a homodimer of 29-kDa (GST-29) subunits and a heterodimer of 29- and 27-kDa (GST-27) subunits, while GST I is twice as active with 1-chloro-2,4-dinitrobenzene as GST II, GST II is about seven times more active against the herbicide, alachlor. Western blotting using antisera raised against GST-29 and GST-27 showed that GST-29 is present throughout the maize plant prior to safener treatment. In contrast, GST-27 is only present in roots of untreated plants but is induced in all the major aerial organs of maize after root-drenching with safener. The amino-acid sequences of proteolytic fragments of GST-27 show that it is related to GST-29 and identical to the 27-kDa subunit of GST IV.
Product Description
Supelco
Dichlormid, PESTANAL®, analytical standard
Pricing and availability is not currently available.