Merck
  • Reactions of 1-bromo-2-[14C]pinacolone with acetylcholinesterase from Torpedo nobiliana. Effects of 5-trimethylammonio-2-pentanone and diisopropyl fluorophosphate.
Reactions of 1-bromo-2-[14C]pinacolone with acetylcholinesterase from Torpedo nobiliana. Effects of 5-trimethylammonio-2-pentanone and diisopropyl fluorophosphate.
Biochimica et biophysica acta (1989-08-31)
S G Cohen, E Salih, M Solomon, S Howard, S B Chishti, J B Cohen
ABSTRACT
1-Bromo-2-[14C]pinacolone, (CH3)3C14COCH2Br [( 14C]BrPin), was prepared from [1-14C]acetyl chloride and tert-butylmagnesium chloride with cuprous chloride catalyst, followed by bromination. It was examined as an active-site directed label for acetylcholinesterase (acetylcholine acetylhydrolase, EC 3.1.1.7) (AcChE). AcChE, isolated from Torpedo nobiliana, has k(cat) = (4.00 +/- 0.04).10(3) s-1, Km = 0.055 +/- 0.008 mM in hydrolysis of acetylthiocholine, and k(cat) = (5.6 +/- 0.2).10(3) s-1, Km = 0.051 +/- 0.003 mM in hydrolysis of acetylcholine. BrPin, binding in the trimethyl cavity, acts initially as a reversible competitive inhibitor, Ki = 0.20 +/- 0.09 mM, and, with time, as an irreversible covalently bound inactivator. Introduction of 14C from [14C]BrPin into Torpedo AcChE at pH 7.0 was followed by SDS-PAGE, autoradiography and scintillation counting, in the absence and presence of 5-trimethylammonio-2-pentanone (TAP), a competitive inhibitor (Ki = 0.075 +/- 0.001 mM) isosteric with acetylcholine; 1.8-1.9 14C was incorporated per inactivated enzyme unit at 50% inactivation. TAP retarded inactivation by [14C]BrPin, and prevented introduction of 0.9-1.1 14C per unit of enzyme protected. Prior inactivation of AcChE by BrPin prevents reaction with [3H]diisopropyl fluorophosphate [( 3H]DFP). Prior inactivation by DFP or [3H]DFP does not prevent reaction with [14C]BrPin, and this subsequent reaction with BrPin does not displace the [3H] moiety. [14C]BrPin alkylates a nucleophile in the active site, and this reaction does not alkylate or utilize the serine-hydroxyl.
Sigma-Aldrich
1-Bromopinacolone, technical grade, ~90%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Bromopinacolone, ≥97%
Pricing and availability is not currently available.