Merck
Inhibition of thioredoxin reductase by lanthanum chloride.
Journal of inorganic biochemistry (2012-10-20)
Anna Citta, Alessandra Folda, Guido Scutari, Luca Cesaro, Alberto Bindoli, Maria Pia Rigobello
ABSTRACT
Lanthanum chloride (LaCl(3)) is a good inhibitor of both the cytosolic and mitochondrial thioredoxin reductase with IC(50) values of 1.75 and 7.46 μM, respectively. On the contrary, the related enzyme glutathione reductase is not inhibited by lanthanum ions even at relatively high concentrations. In the presence of LaCl(3), steady-state kinetics shows a non-competitive type of inhibition with 5,5'-dithiobis-(2-nitrobenzoic acid) (DTNB) as a substrate suggesting no interaction of this ion with the thiol/selenol active site of thioredoxin reductase. Comparison of the electrostatic surface potential of thioredoxin reductase shows that the presence of a trivalent cation such as La(3+) decreases the negatively charged area of the enzyme surface particularly in the region closed to the NADPH binding site. Human ovarian carcinoma cells (A2780 cells) incubated with lanthanum ions show a noticeable inhibition of thioredoxin reductase activity indicating the ability of this ion to reach the active site of the enzyme even in a cellular setting. In addition, A2780 cells treated with LaCl(3) show an increase in reactive oxygen species production in part dependent on thioredoxin reductase inhibition.
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Lanthanum(III) chloride heptahydrate, ACS reagent
Pricing and availability is not currently available.