Merck
  • Design, synthesis, and biological evaluation of novel disubstituted dibenzosuberones as highly potent and selective inhibitors of p38 mitogen activated protein kinase.
Design, synthesis, and biological evaluation of novel disubstituted dibenzosuberones as highly potent and selective inhibitors of p38 mitogen activated protein kinase.
Journal of medicinal chemistry (2012-06-09)
Solveigh C Koeberle, Stefan Fischer, Dieter Schollmeyer, Verena Schattel, Christian Grütter, Daniel Rauh, Stefan A Laufer
ABSTRACT
Synthesis, biological testing, structure-activity relationships (SARs), and selectivity of novel disubstituted dibenzosuberone derivatives as p38 MAP kinase inhibitors are described. Hydrophilic moieties were introduced at the 7-, 8-, and 9-position of the 2-phenylamino-dibenzosuberones, improving physicochemical properties as well as potency. Extremely potent inhibitors were obtained, with half-maximal inhibitory concentration (IC(50)) values in the low nM range in a whole blood assay measuring the inhibition of cytokine release. The high potency of the target compounds together with the outstanding selectivity of this novel class of compounds toward p38 mitogen activated protein (MAP) kinase as compared to other kinases indicate them to be most applicable as tools in pharmacological research and eventually they may foster a new generation of anti-inflammatory drugs.
Product Description
Dibenzosuberone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.