- [Allergic contact eczema to etofenamate and dwarf pine oil].
[Allergic contact eczema to etofenamate and dwarf pine oil].
Sportverletzung Sportschaden : Organ der Gesellschaft fur Orthopadisch-Traumatologische Sportmedizin (1990-06-01)
R Knöll, R Ulrich, W Spallek
PMID2143316
ABSTRACT
A typical case of contact dermatitis following topical applications of etofenamat is reported and a short review of literature is given. Considering the frequent administration of etofenamat, contact sensitization is relatively rare.
Product Description
Etofenamate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Etofenamate for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.