Merck
  • Reduction of potassium currents and phosphatidylinositol 3-kinase-dependent AKT phosphorylation by tumor necrosis factor-(alpha) rescues axotomized retinal ganglion cells from retrograde cell death in vivo.
Reduction of potassium currents and phosphatidylinositol 3-kinase-dependent AKT phosphorylation by tumor necrosis factor-(alpha) rescues axotomized retinal ganglion cells from retrograde cell death in vivo.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2001-03-14)
R Diem, R Meyer, J H Weishaupt, M Bahr
ABSTRACT
Tumor-necrosis-factor-alpha (TNF-alpha) prevented secondary death of retinal ganglion cells (RGCs) after axotomy of the optic nerve in vivo. This RGC rescue was confirmed in vitro in a mixed retinal culture model. In accordance with our previous findings, TNF-alpha decreased outward potassium currents in RGCs. Antagonism of the TNF-alpha-induced decrease in outward potassium currents with the potassium channel opener minoxidilsulfate (as verified by electrophysiology) abolished neuroprotection. Western blot analysis revealed an upregulation of phospho-Akt as a consequence of TNF-alpha-induced potassium current reduction. Inhibition of the phosphatidylinositol 3-kinase-Akt pathway with wortmannin decreased TNF-alpha-promoted RGC survival. These data point to a functionally relevant cytokine-dependent neuroprotective signaling cascade in adult CNS neurons.
Sigma-Aldrich
Minoxidil Sulfate
Pricing and availability is not currently available.