Merck
  • A regulatory role for Staphylococcus aureus toxin-antitoxin system PemIKSa.
A regulatory role for Staphylococcus aureus toxin-antitoxin system PemIKSa.
Nature communications (2013-06-19)
Michal Bukowski, Robert Lyzen, Weronika M Helbin, Emilia Bonar, Agnieszka Szalewska-Palasz, Grzegorz Wegrzyn, Grzegorz Dubin, Adam Dubin, Benedykt Wladyka
ABSTRACT
Toxin-antitoxin systems were shown to be involved in plasmid maintenance when they were initially discovered, but other roles have been demonstrated since. Here we identify and characterize a novel toxin-antitoxin system (pemIKSa) located on Staphylococcus aureus plasmid pCH91. The toxin (PemKSa) is a sequence-specific endoribonuclease recognizing the tetrad sequence U↓AUU, and the antitoxin (PemISa) inhibits toxin activity by physical interaction. Although the toxin-antitoxin system is responsible for stable plasmid maintenance our data suggest the participation of pemIKSa in global regulation of staphylococcal virulence by alteration of the translation of large pools of genes. We propose a common mechanism of reversible activation of toxin-antitoxin systems based on antitoxin transcript resistance to toxin cleavage. Elucidation of this mechanism is particularly interesting because reversible activation is a prerequisite for the proposed general regulatory role of toxin-antitoxin systems.
Sigma-Aldrich
On-Column DNase I Digestion Set, sufficient for 70 preparations
Pricing and availability is not currently available.
Sigma-Aldrich
On-Column DNase I Digestion Set, sufficient for 10 preparations
Pricing and availability is not currently available.