Merck
  • Highly sensitive and specific high-performance liquid chromatographic analysis of 7-hydroxywarfarin, a marker for human cytochrome P-4502C9 activity.
Highly sensitive and specific high-performance liquid chromatographic analysis of 7-hydroxywarfarin, a marker for human cytochrome P-4502C9 activity.
Journal of chromatography. B, Biomedical applications (1995-10-20)
D Lang, R Böcker
ABSTRACT
The formation of 7-hydroxywarfarin in incubations of (S)-warfarin with human liver microsomes reflects their cytochrome P-4502C9 activity. This paper describes a rapid high-performance liquid chromatographic method for the determination of 7-hydroxywarfarin with high sensitivity, selectivity, and a simple sample clean-up procedure. Separation was achieved with a C18 reversed-phase column and quantification by fluorometric detection. The method employs an internal standard resulting in good accuracy and precision. The limit of detection is 150 fmol for 7-hydroxywarfarin.
Sigma-Aldrich
7-Hydroxywarfarin
Pricing and availability is not currently available.