Merck
  • Activated alpha-alkyl-alpha-arylacetic acid enantiomers for stereoselective thin-layer chromatographic and high-performance liquid chromatographic determination of chiral amines.
Activated alpha-alkyl-alpha-arylacetic acid enantiomers for stereoselective thin-layer chromatographic and high-performance liquid chromatographic determination of chiral amines.
Journal of chromatography (1984-04-13)
H Weber, H Spahn, E Mutschler, W Möhrke
ABSTRACT
The separation of racemic benoxaprofen into the two benoxaprofen enantiomers by preparative high-performance liquid chromatography and the application of the activated enantiomers as derivatization reagents for the simultaneous stereoselective determination of chiral amines in biological material is described. Activated (+)- and (-)-benoxaprofen are both shown to be very sensitive and stable chiral fluorescence markers, applicable to thin-layer chromatography as well as to high-performance liquid chromatography.
Sigma-Aldrich
N-Benzylmethylamine, 97%
Pricing and availability is not currently available.