Merck
  • Contact dermatitis from the epoxy resins tetraglycidyl-4,4'-methylene dianiline and o-diglycidyl phthalate in composite material.
Contact dermatitis from the epoxy resins tetraglycidyl-4,4'-methylene dianiline and o-diglycidyl phthalate in composite material.
Contact dermatitis (1984-08-01)
D Burrows, S Fregert, H Campbell, L Trulsson
ABSTRACT
An outbreak of dermatitis occurred in an aircraft factory using epoxy resin composite material. Of 25 operatives, 14 gave positive patch test reactions to the composite material and/or diglycidylether of bisphenol A (DGEBA), tetraglycidyl-4,4'-methylene dianiline (TGMDA), and o-diglycidyl phthalate. This report seems to be the first to demonstrate contact allergy to the two last mentioned epoxy resins. The diglycidylether of bisphenol A used in routine test series picked up only 3 cases of 12 tested.
Sigma-Aldrich
4,4′-Methylenebis(N,N-diglycidylaniline)
Pricing and availability is not currently available.