Merck
  • Presumptive identification of group D streptococci: the bile-esculin test.
Presumptive identification of group D streptococci: the bile-esculin test.
Applied microbiology (1970-08-01)
R R Facklam, M D Moody
ABSTRACT
Six tests commonly used for the presumptive identification of group D streptococci were evaluated. Strains tested included 282 group D streptococci and 366 non-group D. Ratios of percentages of group D to non-group D strains which gave positive reactions for each test are as follows: bile-esculin, 100:2; salt tolerance, 88:24; heat tolerance, 100:80; SF broth, 86:1; KF broth, 99:40; and methylene blue milk reduction, 90:17. These data indicate that the bile-esculin test provided a reliable means of identifying group D streptococci and differentiating them from non-group D streptococci. Methodology for reading and interpreting positive reactions and time of incubation of the bile-esculin medium was defined. Evidence of the need for standardization of salt and heat-tolerance tests was obtained.
Product Description
Millipore
Bile esculin agar, suitable for microbiology, NutriSelect® Plus
Pricing and availability is not currently available.