Merck
  • The relative roles of charge and a recognition peptide in luminal targeting of colorectal cancer by fluorescent polyacrylamide.
The relative roles of charge and a recognition peptide in luminal targeting of colorectal cancer by fluorescent polyacrylamide.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2012-10-02)
Meital Bloch, Yossi Kam, Eylon Yavin, Dorit Moradov, Aviram Nissan, Ilana Ariel, Abraham Rubinstein
ABSTRACT
Real time detection of biomarkers at the mucosal level is imperative for the prevention and efficient treatment of colorectal cancer. Cationized polyacrylamide (CPAA) with increasing charge densities was prepared by radical polymerization of acrylamide and different mol% ratios of N-acryloyl, N'-(tert-butyl-carbonyl) diaminoethane. The NIR fluorophore derivative of IR-783, IR-783-S-Ph-COOH, was attached to the CPAA to give CPAA-783. After selecting the optimal IR-783-S-Ph-COOH ratio that avoids quenching, the preferential binding of the polymer was tested in SW-620, SW-480, HT-29, and LS-174T cancer cells. The optimal polymeric product was tested in situ in gut sac preparations of the dimethylhydrazine induced rat model. To increase the detection capabilities of CPAA-783, the FITC-labeled peptide EPPT1, that targets the cell transmembrane underglycosylated MUC-1 (uMUC-1), was conjugated to the polymer to obtain CPAA-783-EPPT1. The dually labeled modified polymer was tested in HT-29 and LS-174T cells (over expressing uMUC-1), followed by an examination in an orthotopic mouse model. CPAA-783 preferentially bound to the cancer cells, depending on CRC staging. The best binding occurred when the fraction of the cationic monomer was 100 mol%, labeled with 0.75 mol% of IR-783-S-Ph-COOH. An increase in the recognition of the dually labeled polymeric product, CPAA-783-EPPT1, towards HT-29 and LS-174T cells occurred in the lowest EPPT1molar ratio (0.63 mol%) only, probably due to quenching phenomena and steric hindrance. Similar observation was obtained in the orthotopic mice. It is concluded that fluorescently tagged CPAA can be used for the detection of malignant tissues in colorectal cancer after luminal instillation. Dually targeted CPAA with EPPT1is feasible, but requires further optimization.
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Pricing and availability is not currently available.
Sigma-Aldrich
Polyacrylamide, average Mn 150,000
Pricing and availability is not currently available.