Merck
  • Bioactive components in the fruits of Ziziphus jujuba Mill. against the inflammatory irritant action of Euphorbia plants.
Bioactive components in the fruits of Ziziphus jujuba Mill. against the inflammatory irritant action of Euphorbia plants.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2011-10-11)
L Yu, B P Jiang, D Luo, X C Shen, S Guo, J A Duan, Y P Tang
ABSTRACT
Chinese jujube (also known as Chinese date) is the fruit of Ziziphus jujuba Mill. (Rhamnaceae). As a famous folk medicine, it is used as antidote in traditional Chinese formula, Shi Zao Decoction, to relieve the drastic inflammatory irritant nature of Euphorbia species. The irritant activities may cause serious adverse effects in clinical practices. This study aimed to investigate the active components of Z. jujuba through the inhibitory effects on the inflammatory cells activated by Euphorbia kansui and prostratin, a phorbol ester isolated from Euphorbia fischeriana. Peritoneal macrophage of rat and splenic lymphocyte (splenocyte) of mouse were selected to evaluate these actions in vitro. Nitric oxide (NO) release of macrophage and the proliferation of splenocyte were examined through Griess method and MTT assay. TNF-α, as an important pro-inflammatory cytokines, was detected with enzyme-linked immunosorbent assay (ELISA) method. Six fractions extracted from Z. jujuba were evaluated and fraction F (triterpene acids fraction) was demonstrated to be the most active part, and then, 21 compounds isolated from Z. jujuba were tested at the concentrations range from 1 μg/ml to 100 μg/ml. The results show that 7 compounds of them are likely to be active compounds concerning to their pronounced inhibitory action on the activated inflammatory cells. These effects might be helpful to attenuate the irritant action of Euphorbiaceae plants and protect the gastrointestinal tissue from potent inflammatory injury, which should be beneficial to some diseases, like inflammatory bowel disease.
Sigma-Aldrich
Prostratin, ≥98% (HPLC)
Pricing and availability is not currently available.