Merck
  • Binding and inhibition of human spermidine synthase by decarboxylated S-adenosylhomocysteine.
Binding and inhibition of human spermidine synthase by decarboxylated S-adenosylhomocysteine.
Protein science : a publication of the Protein Society (2011-09-08)
Jolita Sečkutė, Diane E McCloskey, H Jeanette Thomas, John A Secrist, Anthony E Pegg, Steven E Ealick
ABSTRACT
Aminopropyltransferases are essential enzymes that form polyamines in eukaryotic and most prokaryotic cells. Spermidine synthase (SpdS) is one of the most well-studied enzymes in this biosynthetic pathway. The enzyme uses decarboxylated S-adenosylmethionine and a short-chain polyamine (putrescine) to make a medium-chain polyamine (spermidine) and 5'-deoxy-5'-methylthioadenosine as a byproduct. Here, we report a new spermidine synthase inhibitor, decarboxylated S-adenosylhomocysteine (dcSAH). The inhibitor was synthesized, and dose-dependent inhibition of human, Thermatoga maritima, and Plasmodium falciparum spermidine synthases, as well as functionally homologous human spermine synthase, was determined. The human SpdS/dcSAH complex structure was determined by X-ray crystallography at 2.0 Å resolution and showed consistent active site positioning and coordination with previously known structures. Isothermal calorimetry binding assays confirmed inhibitor binding to human SpdS with K(d) of 1.1 ± 0.3 μM in the absence of putrescine and 3.2 ± 0.1 μM in the presence of putrescine. These results indicate a potential for further inhibitor development based on the dcSAH scaffold.
Sigma-Aldrich
S-(5′-Adenosyl)-3-thiopropylamine, ≥98.0% (HPLC)
Pricing and availability is not currently available.