- Fire-eater's pneumonia: two case reports of accidentally aspirated paraffin oil.
Fire-eater's pneumonia: two case reports of accidentally aspirated paraffin oil.
Fire-eater's pneumonia is a chemical pneumonitis that can develop after accidental aspiration of liquid hydrocarbon-based fuel during a flame-blowing or a fire-eating performance. Typical findings of the patient are similar with any infectious pneumonia: chest pain, shortness of breath, cough, fever, and hemoptysis can be seen. We report two cases of acute paraffin oil-induced pneumonia due to accidental aspiration during fire-eating performance. The symptoms and course of respiratory manifestations and the treatment strategies of fire-eater's pneumonia are reviewed.