Merck
  • Blockade of LTB4-induced chemotaxis by bioactive molecules interfering with the BLT2-Galphai interaction.
Blockade of LTB4-induced chemotaxis by bioactive molecules interfering with the BLT2-Galphai interaction.
Biochemical pharmacology (2010-01-26)
Joo-Young Kim, Won-Kyu Lee, Yeon Gyu Yu, Jae-Hong Kim
ABSTRACT
BLT2, a low-affinity leukotriene B4 (LTB4) receptor, is a member of the G-protein coupled receptor (GPCR) family and is involved in the pathogenesis of inflammatory diseases such as asthma. Despite its clinical implications, however, no pharmacological inhibitors are available. In the present study, we screened for small molecules that interfere with the interaction between the third intracellular loop region of BLT2 (BLT2iL3) and the Galphai3 protein subunit (Galphai3), using a high-throughput screening (HTS) assay with a library of 1040 FDA-approved drugs and bioactive compounds. We identified two small molecules-purpurin [1,2,4-trihydroxy-9,10-anthraquinone; IC50 = 1.6 microM for BLT2] and chloranil [tetrachloro-1,4-benzoquinone; IC50 = 0.42 microM for BLT2]-as specific BLT2-blocking agents. We found that blockade of the BLT2iL3-Galphai3 interaction by these small molecules inhibited the BLT2-downstream signaling cascade. For example, BLT2-signaling to phosphoinositide-3 kinase (PI3K)/Akt phosphorylation was completely abolished by these molecules. Furthermore, we observed that these small molecules blocked LTB4-induced chemotaxis by inhibiting the BLT2-PI3K/Akt-downstream, Rac1-reactive oxygen species-dependent pathway. Taken together, our results show that purpurin and chloranil interfere with the interaction between BLT2iL3 and Galphai3 and thus block the biological functions of BLT2 (e.g., chemotaxis). The present findings suggest a potential application of purpurin and chloranil as pharmacological therapeutic agents against BLT2-associated inflammatory human diseases.
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%
Pricing and availability is not currently available.
Supelco
Chloranil, PESTANAL®, analytical standard
Pricing and availability is not currently available.