Merck
Evaluation of leaching behavior of pendimethalin in sandy loam soil.
Environmental monitoring and assessment (2009-01-09)
Indu Chopra, Beena Kumari, S K Sharma
ABSTRACT
The mobility of pendimethalin in sandy loam soil was studied in soil columns under laboratory conditions at two application rates, 1.0 and 2.0 kg a.i. ha(-1), with simulated rainfall of 300 mm. The maximum concentration of the herbicide was found in the top 10 cm layer, though it was found distributed in soil at all the depths at both the doses.
Product Description
Supelco
Pendimethalin, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Pendimethalin solution, 100 μg/mL in methanol, PESTANAL®, analytical standard
Pricing and availability is not currently available.