Merck
  • Non-covalent binding of azo compound to peptide chain: interactions of biebrich scarlet and naphthochrome green with four model proteins.
Non-covalent binding of azo compound to peptide chain: interactions of biebrich scarlet and naphthochrome green with four model proteins.
Amino acids (2008-03-28)
Hong-Wen Gao, Xiang-Hu Liu, Zhi Qiu, Lu Tan
ABSTRACT
We studied the non-specific interactions of two azo compounds: biebrich scarlet (BS) and naphthochrome green (NG), with four model proteins: bovine serum albumin, ovalbumin, poly-L-lysine and hemoglobin by UV-VIS spectrometry, fluorophotometry and circular dichroism melting technique. The optimal acidities of NG and BS for binding to proteins correspond to the physiological pHs of skin and gastro tissues. The saturation binding numbers of BS and NG on peptide chains were determined and the effects of electrolytes and temperature were investigated. These interactions were fitted by the Temkin absorption model and their thermodynamic parameters were calculated. The different bindings of BS and NG to proteins were compared from their molecular structures. We inferred that an ion-pair electrostatic interaction first fixes azo compounds to basic amino acid residues and subsequent binding involves the collective action of other non-covalent bonds: hydrogen bond, van der Waals force, and hydrophobic interaction. This combination of bonds caused a change of secondary conformation of protein from beta-sheet to helix and the possible process was illustrated. The potential protein toxicity resulting from such a non-specific binding was analyzed. Besides, the interaction of BS with peptide chains was applied to protein assay.
Sigma-Aldrich
Ponceau BS, Dye content ≥55 %
Pricing and availability is not currently available.