Merck
On the synthesis of protopine alkaloids.
The Journal of organic chemistry (2007-08-21)
Yasuhiro Wada, Harumi Kaga, Shiho Uchiito, Eri Kumazawa, Miho Tomiki, Yu Onozaki, Nobuhito Kurono, Masao Tokuda, Takeshi Ohkuma, Kazuhiko Orito
ABSTRACT
For the synthesis of protopine alkaloids, we studied a reaction sequence based on a ring enlargement of indeno[2,1-a][3]benzazepines by a singlet oxygen oxygenation, followed by conversion of an amide carbonyl group of the resultant 10-membered keto-lactam to a methylene group, which is the last step for completion of the synthesis. The key substances, indeno[2,1-a][3]benzazepines, were prepared by Bischler-Napieralski cyclization of alkoxy-substituted 1-(2-bromobenzyl)-3-benzazepin-2-ones. Steric effects of the substituents in this synthesis were examined.
Sigma-Aldrich
Protopine hydrochloride, ≥98%, solid
Pricing and availability is not currently available.