Merck
Multifunctional polymeric nanoparticles from diverse bioactive agents.
Journal of the American Chemical Society (2006-03-30)
Paul A Bertin, Julianne M Gibbs, Clifton Kwang-Fu Shen, C Shad Thaxton, William A Russin, Chad A Mirkin, Sonbinh T Nguyen
ABSTRACT
We present a rational approach for assembling diverse bioactive agents, such as DNA, proteins, and drug molecules, into core-shell multifunctional polymeric nanoparticles (PNPs) that can be internalized in human breast cancer cells. Using ring-opening metathesis polymerization (ROMP), block copolymers containing small-molecule drug segments (>50% w/w) and tosylated hexaethylene glycol segments were prepared and assembled into PNPs that allowed for the surface conjugation of single-stranded DNA sequences and/or tumor-targeting antibodies. The resulting antibody-functionalized particles were readily uptaken by breast cancer cells that overexpressed the corresponding antigens.
Sigma-Aldrich
Hexaethylene glycol, 97%
Pricing and availability is not currently available.