Merck
Synthesis and properties of a new type DNA dendrimer.
Nucleic acids symposium series (2003-08-09)
Y Suzuki, T Otomo, H Ozaki, H Sawai
ABSTRACT
We have designed a new type of a DNA dendrimer which has rigid branched structure. The branching molecule was prepared from 1,3,5-tribromobenzene. The dendrimer unit, in which three oligonucleotide-chains, two molecules of T15 and one molecule of A15, linked to the branching molecule, was synthesized by an automated DNA synthesizer. The properties of the dendrimer unit and dendrimer formation by inter-molecular association of T15 and A15 chains of the dendrimer unit will be presented.
Sigma-Aldrich
1,3,5-Tribromobenzene, 98%
Pricing and availability is not currently available.