Merck
(2-Naphthoxy)acetyl chloride, a simple fluorescent reagent.
Journal of chromatography. A (2003-03-05)
Tsai-Hui Duh, Hsin-Lung Wu, Hwang-Shang Kou, Chi-Yu Lu
ABSTRACT
In continuing the search for fluorescent reagents for analytical derivatization in chromatography, we found a simple chemical, (2-naphthoxy)acetyl chloride, with potential fluorophore/chromophore characteristics for the highly sensitive detection of analytes with an amino function. The reagent has an auxochrome (a substituted alkoxy moiety) attached to the fluorophoric/chromophoric naphthalene system, resulting in favorable spectrophotometric properties. The reagent can be easily prepared from (2-naphthoxy)acetic acid and has been used in organic synthesis; it is initially introduced as a fluorescent reagent to derivatise amantadine and memantine (amino pharmaceuticals) as model analytes. The resulting naphthoxy derivatives of the drugs can be analyzed at sub-microM levels by HPLC with fluorimetric detection (excitation wavelength 227 nm, emission wavelength 348 nm). Application of the reagent to the fluorimetric derivatization of important biological amines for sensitive detection can be expected.
Sigma-Aldrich
2-Naphthoxyacetic acid, BioReagent, suitable for plant cell culture, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
2-Naphthoxyacetic acid, 98%
Pricing and availability is not currently available.