Merck
Oxidation of phenols by laccase and laccase-mediator systems.
European journal of biochemistry (2002-10-24)
Francesca D'Acunzo, Carlo Galli, Bernardo Masci
ABSTRACT
To investigate how solubility and steric issues affect the laccase-catalysed oxidation of phenols, a series of oligomeric polyphenol compounds, having increasing size and decreasing solubility in water, was incubated with laccase. The extent of substrate conversion, and the nature of the products formed in buffered aqueous solutions, were compared to those obtained in the presence of an organic cosolvent, and also in the presence of two mediating species, i.e. N-hydroxyphthalimide (HPI) and 2,2,6,6-tetramethylpiperidin-1-yloxy (TEMPO). This approach showed not only an obvious role of solubility, but also a significant role of the dimension of the substrate upon the enzymatic reactivity. In fact, reactivity decreases as substrate size increases even when solubility is enhanced by a cosolvent. This effect may be ascribed to limited accessibility of encumbered substrates to the enzyme active site, and can be compensated through the use of the appropriate mediator. While TEMPO was highly efficient at enhancing the reactivity of large, less soluble substrates, HPI proved less effective. In addition, whereas the laccase/HPI system afforded the same products as laccase alone, the use of TEMPO provided a different product with high specificity. These results offer the first evidence of the role of 'oxidation shuttles' that the mediators of laccase may have, but also suggest two promising routes towards an environmentally friendly process for kraft pulp bleaching: (a) the identification of mediators which, once oxidized by laccase, are able to target strategic functional groups present in lignin, and (b) the introduction of those strategic functional groups in an appropriate pretreatment.
Sigma-Aldrich
N-Hydroxyphthalimide, 97%
Pricing and availability is not currently available.