Merck
  • The use of (oligonucleotide-maleic anhydride copolymer) conjugates in nucleic acid diagnostic assays: effect of the number of oligonucleotides per polymer on test sensitivity improvement.
The use of (oligonucleotide-maleic anhydride copolymer) conjugates in nucleic acid diagnostic assays: effect of the number of oligonucleotides per polymer on test sensitivity improvement.
Nucleosides, nucleotides & nucleic acids (2001-09-21)
C Minard-Basquin, C Chaix, C Pichot
ABSTRACT
(Polymer-oligonucleotide) conjugates were obtained via direct ON synthesis from the poly(ethylene-alt-maleic anhydride: PEMA) grafted onto glass bead surfaces. The effect of the ON number per polymer chain on binding was evaluated through Tm experiments. Results were correlated with ELOSA (Enzyme Linked Oligosorbent Assay) tests run with conjugates in the capture step.
Sigma-Aldrich
Poly(ethylene-alt-maleic anhydride), average Mw 100,000-500,000, powder
Pricing and availability is not currently available.