Merck
  • Human sweat and 2-oxopentanoic acid elicit a landing response from Anopheles gambiae.
Human sweat and 2-oxopentanoic acid elicit a landing response from Anopheles gambiae.
Medical and veterinary entomology (2000-06-29)
T P Healy, M J Copland
ABSTRACT
A wind tunnel bioassay and video to observe mosquitoes landing on heated glass cylinders were used to test sweat and some derivatives for responses of Anopheles gambiae Giles (Diptera: Culicidae), a highly anthropophilic African species of malaria vector. Filter papers impregnated with human sweat and a diethyl ether extract from the filter papers elicited significantly more landings than a water control (P<0.001). The concentration of lactic acid in the extract was determined by GLC assay, but bioassays of an equivalent dose of lactic acid (from a commercial supplier) did not elicit landings. Chemical analysis of the extract by combined GLC/mass spectrometry indicated the presence of 73 compounds, of which 40 were tentatively identified. The major components of the extract were aliphatic carboxylic acids. An artificial blend of 22 carboxylic acids did not elicit landings. Bioassays of 2-oxopentanoic acid elicited significantly more, landings (P<0.001). The possible importance of oxo-carboxylic acids for host-seeking by anthropophilic mosquitoes is discussed and their use for trapping is suggested.
Sigma-Aldrich
2-Oxovaleric acid, ≥98.0% (T)
Pricing and availability is not currently available.