Merck
  • Determination of 3,3'-dichlorobenzidine and its degradation products in environmental samples with a small low-field Fourier transform ion cyclotron resonance mass spectrometer.
Determination of 3,3'-dichlorobenzidine and its degradation products in environmental samples with a small low-field Fourier transform ion cyclotron resonance mass spectrometer.
Journal of the American Society for Mass Spectrometry (1999-10-28)
M C Nyman, J Pérez, E R Blatchley, H I Kenttämaa
ABSTRACT
3,3'-Dichlorobenzidine (DCB) and its degradation products, 3-chlorobenzidine (MCB) and benzidine, are of environmental concern because of their carcinogenic nature. The suitability of a small Fourier transform ion cyclotron resonance (FT-ICR) mass spectrometer for the analysis of these environmental contaminants in different types of matrices was explored. All the measurements were carried out by depositing the sample solution directly on a disk that was introduced into the mass spectrometer. This approach is very fast and simple because it requires no prior chromatographic separation or derivatization. Calibration curves determined by collecting 70-eV electron ionization mass spectra of neat samples yielded lower limits of detection of 29 and 17 pg (total amount on the solids probe) for DCB and benzidine, respectively (based on a signal to noise ratio of > or = 2:1), while chemical ionization with ammonia resulted in lower limits of detection of 21 pg for DCB and 9 pg for benzidine (total amount on the solids probe). FT-ICR analysis of sediments collected from Lake Macatawa (Holland, MI) verified the presence of DCB in this complex, environmentally significant sample matrix. Laboratory experiments designed to probe biodegradation and photodegradation pathways showed that DCB undergoes sequential dehalogenation to yield MCB and then benzidine under exposure to microorganisms and under simulated tropospheric solar radiation. The ability of the FT-ICR to determine elemental compositions of compounds introduced as described above was demonstrated for one of the degradation products.
Product Description
Supelco
3,3′-Dichlorobenzidine, analytical standard
Pricing and availability is not currently available.