Merck
  • Enhanced alpha-ketoglutaric acid production in Yarrowia lipolytica WSH-Z06 by regulation of the pyruvate carboxylation pathway.
Enhanced alpha-ketoglutaric acid production in Yarrowia lipolytica WSH-Z06 by regulation of the pyruvate carboxylation pathway.
Applied microbiology and biotechnology (2012-06-09)
Xiaoxia Yin, Catherine Madzak, Guocheng Du, Jingwen Zhou, Jian Chen
ABSTRACT
In previous research, a thiamine-auxotrophic yeast for alpha-ketoglutaric acid (KGA) overproduction was screened in our laboratory and named Yarrowia lipolytica WSH-Z06 (CCTCC no. M207143). However, the high concentration of by-products (mainly pyruvate) limited its application on an industrial scale. To enhance KGA production and reduce pyruvate (PA) accumulation, the pyruvate carboxylation pathway was regulated. By overexpressing the pyruvate carboxylase genes ScPYC1 from Saccharomyces cerevisiae and RoPYC2 from Rhizopus oryzae in Y. lipolytica WSH-Z06, the yields of KGA in Y. lipolytica-ScPYC1 and Y. lipolytica-RoPYC2 increased by 24.5 and 35.3 %, and the yields of PA decreased by 51.9 and 69.8 % in shake flasks, respectively. These changes in the expression levels and activities of key intracellular enzymes showed that enhancing the pyruvate carboxylation pathway had successfully redistributed the carbon flux from PA to KGA. Finally, by controlling the pH in a 3-L fermenter, the maximum concentration of KGA in Y. lipolytica-RoPYC2 reached 62.5 g L⁻¹ with an evident decrease in PA yield from 35.2 to 13.5 g L⁻¹.
Sigma-Aldrich
Pyruvate Carboxylase from bovine liver, buffered aqueous glycerol solution, 5-25 units/mg protein (BCA)
Pricing and availability is not currently available.