Merck
  • Plantain and banana starches: granule structural characteristics explain the differences in their starch degradation patterns.
Plantain and banana starches: granule structural characteristics explain the differences in their starch degradation patterns.
Journal of agricultural and food chemistry (2011-05-20)
Claudinéia Aparecida Soares, Fernanda Helena Gonçalves Peroni-Okita, Mateus Borba Cardoso, Renata Shitakubo, Franco Maria Lajolo, Beatriz Rosana Cordenunsi
ABSTRACT
Different banana cultivars were used to investigate the influences of starch granule structure and hydrolases on degradation. The highest degrees of starch degradation were observed in dessert bananas during ripening. Scanning electron microscopy images revealed smooth granule surface in the green stage in all cultivars, except for Mysore. The small and round granules were preferentially degraded in all of the cultivars. Terra demonstrated a higher degree of crystallinity and a short amylopectin chain length distribution, resulting in high starch content in the ripe stage. Amylose content and the crystallinity index were more strongly correlated than the distribution of amylopectin branch chain lengths in banana starches. α- and β-amylase activities were found in both forms, soluble in the pulp and associated with the starch granule. Starch-phosphorylase was not found in Mysore. On the basis of the profile of α-amylase in vitro digestion and the structural characteristics, it could be concluded that the starch of plantains has an arrangement of granules more resistant to enzymes than the starch of dessert bananas.
Sigma-Aldrich
β-Amylase from sweet potato
Pricing and availability is not currently available.
Sigma-Aldrich
β-Amylase from sweet potato, Type I-B, ammonium sulfate suspension, ≥750 units/mg protein (E1%/280)
Pricing and availability is not currently available.
Sigma-Aldrich
β-Amylase from barley, Type II-B, 20-80 units/mg protein (biuret)
Pricing and availability is not currently available.