Merck
  • Diminished iron concentrations increase adenosine A(2A) receptor levels in mouse striatum and cultured human neuroblastoma cells.
Diminished iron concentrations increase adenosine A(2A) receptor levels in mouse striatum and cultured human neuroblastoma cells.
Experimental neurology (2008-11-18)
Seema Gulyani, Christopher J Earley, Simonetta Camandola, Stuart Maudsley, Sergi Ferré, Mohamed R Mughal, Bronwen Martin, Aiwu Cheng, Marc Gleichmann, Byron C Jones, Richard P Allen, Mark P Mattson
ABSTRACT
Brain iron insufficiency has been implicated in several neurological disorders. The dopamine system is consistently altered in studies of iron deficiency in rodent models. Changes in striatal dopamine D(2) receptors are directly proportional to the degree of iron deficiency. In light of the unknown mechanism for the iron deficiency-dopamine connection and because of the known interplay between adenosinergic and dopaminergic systems in the striatum we examined the effects of iron deficiency on the adenosine system. We first attempted to assess whether there is a functional change in the levels of adenosine receptors in response to this low iron. Mice made iron-deficient by diet had an increase in the density of striatal adenosine A(2A) (A(2A)R) but not A(1) receptor (A(1)R) compared to mice on a normal diet. Between two inbred murine strains, which had 2-fold differences in their striatal iron concentrations under normal dietary conditions, the strain with the lower striatal iron had the highest striatal A(2A)R density. Treatment of SH-SY5Y (human neuroblastoma) cells with an iron chelator resulted in increased density of A(2A)R. In these cells, A(2A)R agonist-induced cyclic AMP production was enhanced in response to iron chelation, also demonstrating a functional upregulation of A(2A)R. A significant correlation (r(2)=0.79) was found between a primary marker of cellular iron status (transferrin receptor (TfR)) and A(2A)R protein density. In conclusion, the A(2A)R is increased across different iron-insufficient conditions. The relation between A(2A)R and cellular iron status may be an important pathway by which adenosine may alter the function of the dopaminergic system.
Sigma-Aldrich
CGS-21680 hydrochloride hydrate, solid
Pricing and availability is not currently available.