- Bioprosthetic Valve Deterioration: Accumulation of Circulating Proteins and Macrophages in the Valve Interstitium.
Bioprosthetic Valve Deterioration: Accumulation of Circulating Proteins and Macrophages in the Valve Interstitium.
Histologic evaluations revealed excessive accumulations of macrophages and absence of fibroblastic interstitial cells in explanted bioprosthetic valves. Comprehensive gene and protein expression analysis and histology unveiled an accumulation of fibrinogen and plasminogen, an activator of infiltrated macrophages, from degenerated valve surfaces in the interstitial spaces. These pathologies were completely reproduced in a goat model replaced with an autologous pericardium-derived aortic valve. Further preclinical animal experiments using goats demonstrated that preventing infiltration of macrophages and circulating proteins by increasing collagen density and leaflet strength is an effective treatment option.