Merck
  • Losses of Yeast-Fermented Carbon Dioxide during Prolonged Champagne Aging: Yes, the Bottle Size Does Matter!
Losses of Yeast-Fermented Carbon Dioxide during Prolonged Champagne Aging: Yes, the Bottle Size Does Matter!
ACS omega (2023-07-03)
Gérard Liger-Belair, Chloé Khenniche, Clara Poteau, Carine Bailleul, Virginie Thollin, Clara Cilindre
ABSTRACT
When it comes to champagne tasting, dissolved CO2 is a key compound responsible for the very much sought-after effervescence in glasses. Nevertheless, the slow decrease of dissolved CO2 during prolonged aging of the most prestigious cuvees raises the issue of how long champagne can age before it becomes unable to form CO2 bubbles during tasting. Measurements of dissolved CO2 concentrations were done on a collection of 13 successive champagne vintages stored in standard 75 cL bottles and 150 cL magnums showing prolonged aging ranging from 25 to 47 years. The vintages elaborated in magnums were found to retain their dissolved CO2 much more efficiently during prolonged aging than the same vintages elaborated in standard bottles. A multivariable exponential decay-type model was proposed for the theoretical time-dependent concentration of dissolved CO2 and the subsequent CO2 pressure in the sealed bottles during champagne aging. The CO2 mass transfer coefficient through the crown caps used to seal champagne bottles prior to the 2000s was thus approached in situ with a global average value of K ≈ 7 × 10-13 m3 s-1. Moreover, the shelf-life of a champagne bottle was examined in view of its ability to still produce CO2 bubbles in a tasting glass. A formula was proposed to estimate the shelf-life of a bottle having experienced prolonged aging, which combines the various relevant parameters at play, including the geometric parameters of the bottle. Increasing the bottle size is found to tremendously increase its capacity to preserve dissolved CO2 and therefore the bubbling capacity of champagne during tasting. For the very first time, a long time-series dataset combined with a multivariable model indicates that the bottle size plays a crucial role on the progressive decay of dissolved CO2 experienced by champagne during aging.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Pricing and availability is not currently available.