Merck
  • The CaMKIIα hub ligand Ph-HTBA promotes neuroprotection after focal ischemic stroke by a distinct molecular interaction.
The CaMKIIα hub ligand Ph-HTBA promotes neuroprotection after focal ischemic stroke by a distinct molecular interaction.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2022-10-25)
Nane Griem-Krey, Stine J Gauger, Emma K Gowing, Louise Thiesen, Bente Frølund, Andrew N Clarkson, Petrine Wellendorph
ABSTRACT
Ca2+/calmodulin-dependent protein kinase II alpha (CaMKIIα) is a potential target for acute neuroprotection due to its key role in physiological and pathological glutamate signaling. The hub domain organizes the CaMKII holoenzyme into large oligomers, and additional functional effects on holoenzyme activation have lately emerged. We recently reported that compounds related to the proposed neuromodulator γ-hydroxybutyrate (GHB) selectively bind to the CaMKIIα hub domain and increase hub thermal stabilization, which is believed to have functional consequences and to mediate neuroprotection. However, the detailed molecular mechanism is unknown. In this study, we functionally characterize the novel and brain permeable GHB analog (E)-2-(5-hydroxy-2-phenyl-5,7,8,9-tetrahydro-6H-benzo[7]annulen-6-ylidene)acetic acid (Ph-HTBA). Administration of a single dose of Ph-HTBA at a clinically relevant time point (3-6 h after photothrombotic stroke) promotes neuroprotection with a superior effect at low doses compared to the smaller GHB analog 3-hydroxycyclopent-1-enecarboxylic acid (HOCPCA). In contrast to HOCPCA, Ph-HTBA reduces Ca2+-stimulated CaMKIIα Thr286 autophosphorylation in primary cortical neurons and substrate phosphorylation of recombinant CaMKIIα, potentially contributing to its neuroprotective effect. Supported by previous in silico docking studies, we suggest that Ph-HTBA makes distinct molecular interactions with the hub cavity, which may contribute to its differential functional profile and superior neuroprotective effect compared to HOCPCA. Together, this highlights Ph-HTBA as a promising tool to study hub functionality, but also as a good candidate for clinical development.
Sigma-Aldrich
Calmodulin from bovine testes, BioUltra, ≥98% (SDS-PAGE), lyophilized powder, essentially salt free
Pricing and availability is not currently available.