Merck
  • Treponema denticola chymotrypsin-like protease as associated with HPV-negative oropharyngeal squamous cell carcinoma.
Treponema denticola chymotrypsin-like protease as associated with HPV-negative oropharyngeal squamous cell carcinoma.
British journal of cancer (2018-06-23)
Anna Kaisa Kylmä, Lauri Jouhi, Dyah Listyarifah, Hesham Mohamed, Antti Mäkitie, Satu Maria Remes, Caj Haglund, Timo Atula, Mikko T Nieminen, Timo Sorsa, Jaana Hagström
ABSTRACT
An opportunistic oral pathogen, Treponema denticola (Td), has been linked to orodigestive carcinogenesis, but its role in oropharyngeal squamous cell carcinoma (OPSCC) has remained open. We evaluated the presence of Td chymotrypsin-like protease (Td-CTLP) in a series of 201 unselected consecutive OPSCC patients, and the relation of the Td-CTLP to human papillomavirus (HPV) status, to expression of toll-like receptors (TLR) 5, 7, and 9, and to clinical parameters and patient outcome. Clinicopathological data came from hospital registries. The expression of cell surface-bound Td-CTLP was evaluated by immunohistochemistry. Immunoexpression of TLRs 5, 7, and 9, and HPV status we studied earlier in this patient series. We detected Td-CTLP in 81% of the OPSCC, and especially in HPV-negative tumours (48% of all OPSCCs). Among the HPV-positive tumours (52% of all OPSCCs), low Td-CTLP expression associated with low TLR 5 and high TLR 7 expression. Among those HPV-negative, higher TLR 5 and lower TLR 7 expression associated with high Td-CTLP expression. Strong Td-CTLP expression associated with poor disease-specific survival, but no similar association among HPV-positive and HPV-negative subgroups emerged. Td-CTLP was highly expressed in OPSCC and was associated with the HPV status of tumour tissue.
Product Description
Protein A-Sepharose CL-4B, Cytiva 17-0780-01, pack of 1.5 g
Pricing and availability is not currently available.