Merck
  • Interferon-inducible TRIM22 contributes to maintenance of HIV-1 proviral latency in T cell lines.
Interferon-inducible TRIM22 contributes to maintenance of HIV-1 proviral latency in T cell lines.
Virus research (2019-05-29)
Filippo Turrini, Fabio Saliu, Greta Forlani, Atze T Das, Carine Van Lint, Roberto S Accolla, Ben Berkhout, Guido Poli, Elisa Vicenzi
ABSTRACT
The human immunodeficiency virus type-1 (HIV-1) establishes a state of latent infection in a small number of CD4+ T lymphocytes that, nonetheless, represent a major obstacle to viral eradication. We here show that Tripartite Motif-containing protein 22 (TRIM22), an epigenetic inhibitor of Specificity protein 1 (Sp1)-dependent HIV-1 transcription, is a relevant factor in maintaining a state of repressed HIV-1 expression at least in CD4+ T cell lines. By knocking-down (KD) TRIM22 expression, we observed an accelerated reactivation of a doxycycline (Dox)-controlled HIV-1 replication in the T lymphocytic SupT1 cell line. Furthermore, we here report for the first time that TRIM22 is a crucial factor for maintaining a state of HIV-1 quiescence in chronically infected ACH2 -T cell line while its KD potentiated HIV-1 expression in both ACH-2 and J-Lat 10.6 cell lines upon cell stimulation with either tumor necrosis factor-α (TNF-α) or histone deacetylase inhibitors (HDACi). In conclusion, TRIM22 is a novel determinant of HIV-1 latency, at least in T cell lines, thus representing a potential pharmacological target for strategies aiming at curtailing or silencing the pool of latently infected CD4+ T lymphocytes constituting the HIV-1 reservoir in individuals receiving combination antiretroviral therapy.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-FITC antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-TRIM22 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.