Merck
  • Tissue-type plasminogen activator is a target of the tumor suppressor gene maspin.
Tissue-type plasminogen activator is a target of the tumor suppressor gene maspin.
Proceedings of the National Academy of Sciences of the United States of America (1998-01-22)
S Sheng, B Truong, D Fredrickson, R Wu, A B Pardee, R Sager
ABSTRACT
The maspin protein has tumor suppressor activity in breast and prostate cancers. It inhibits cell motility and invasion in vitro and tumor growth and metastasis in nude mice. Maspin is structurally a member of the serpin (serine protease inhibitors) superfamily but deviates somewhat from classical serpins. We find that single-chain tissue plasminogen activator (sctPA) specifically interacts with the maspin reactive site loop peptide and forms a stable complex with recombinant maspin [rMaspin(i)]. Major effects of rMaspin(i) are observed on plasminogen activation by sctPA. First, rMaspin(i) activates free sctPA. Second, it inhibits sctPA preactivated by poly-D-lysine. Third, rMaspin(i) exerts a biphasic effect on the activity of sctPA preactivated by fibrinogen/gelatin, acting as a competitive inhibitor at low concentrations (< 0.5 microM) and as a stimulator at higher concentrations. Fourth, 38-kDa C-terminal truncated rMaspin(i) further stimulates fibrinogen/gelatin-associated sctPA. rMaspin(i) acts specifically; it does not inhibit urokinase-type plasminogen activator, plasmin, chymotrypsin, trypsin, or elastase. Our kinetic data are quantitatively consistent with a model in which two segregated domains of maspin interact with the catalytic and activating domains of sctPA. These complex interactions between maspin and sctPA in vitro suggest a mechanism by which maspin regulates plasminogen activation by sctPA bound to the epithelial cell surface.
Sigma-Aldrich
Monoclonal Anti-Chicken Egg Albumin (Ovalbumin) antibody produced in mouse, clone OVA-14, ascites fluid
Pricing and availability is not currently available.