Merck
  • Determination of specific activities and kinetic constants of biotinidase and lipoamidase in LEW rat and Lactobacillus casei (Shirota).
Determination of specific activities and kinetic constants of biotinidase and lipoamidase in LEW rat and Lactobacillus casei (Shirota).
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-08-01)
Kou Hayakawa, Lei Guo, Elena A Terentyeva, Xiao-Kang Li, Hiromitsu Kimura, Masahiko Hirano, Kazuyuki Yoshikawa, Takeaki Nagamine, Noriyuki Katsumata, Tsutomu Ogata, Toshiaki Tanaka
ABSTRACT
Enzyme kinetic parameters, such as K(m), V(max) (or V), k(cat)/K(m), and K(i) (by biotin or lipoic acid) for biotinidase and lipoamidase were determined in Lewis (LEW) rat and Lactobacillus casei (Shirota) using fluorimetric high-performance liquid chromatography (HPLC). It was found that the final protein concentration below 0.1mg/ml is sufficient to obtain linear hydrolytic reaction and to determine the Michaelis-Menten type kinetic parameters (K(m), V, K(i)). We applied this HPLC enzyme assay method onto the rat and some bacteria. The highest specific activities (Vs) for biotinidase were found in Lactobacillus casei (Shirota) and rat kidney. It was also found that the largest K(i) by product for biotinidase and lipoamidase were present in the Lactobacillus casei (Shirota). There has been found specie (between rat and mouse) differences and tissue (organ) differences, together with tissue region differences and sex differences in some tissues. Summary of the distributions of both enzymes in LEW rat was also presented. Therefore, this HPLC determination method for the enzyme kinetic parameters in tissues is expected to be an indispensable tool for the investigation of the various diseases in humans.
Sigma-Aldrich
Biotin 4-amidobenzoic acid sodium salt, ≥95%
Pricing and availability is not currently available.